Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution
Catalog # PRLSMC312937.6010
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
About this item
:
:98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589