Eosin B spirit soluble ≥95%, Sigma-Aldrich®
212954-25G
:
- Förpackningstyp:Glass bottle for solids
- Förp.:25 g
- Seq:3308
- Appearance (Colour):Orange to Red-Orange
- Appearance (Form):Powder, Crystals, Crystalline Powder and/or Chunks
- Infrared spectrum:Conforms to Structure
- UV/Vis absorbance:515 - 521
- SD UVVI e518:≥7000
- Carbon:≥35.20%
- Nitrogen:≥3.40%
:
:580.09 g/mol
:12,03638
:275 °C (dec., lit.)
:M-Clarity™ quality level = MQ100
:Room Temperature
:208-943-1
:56360-46-4
:548-24-3
:C20H8Br2N2O9
:MFCD00066762