
Imidazolidinyl urea NF
Supplier: Spectrum Chemical
New Product
Certificates
About this item
CAS Number:39236-46-9
:
Formula:C₁₁H₁₆N₈O₈
MDL Number:MFCD00221482
:388.30 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (As Nitrogen):26.0 - 28.0%
- Color and Clarity of Solution:To Pass Test
- pH of a 1% Solution:6.0 - 7.5
- Loss On Drying:max. 3.0%
- Residue On Ignition:max. 3.0%