Order Entry
Sweden
Orders LinkContactUsLinkComponent
STEMdiff™ Cardiomyocyte Maintenance Kit
STEMdiff™ Cardiomyocyte Maintenance Kit
STEMdiff™ Cardiomyocyte maintenance kit, 1 KIT
Catalog # STMC05020
Marketsource
STEMdiff™ Cardiomyocyte Maintenance Kit
Catalog # STMC05020
Marketsource
CAS Number:  
STEMdiff™ Cardiomyocyte Maintenance Kit
STEMdiff™ Cardiomyocyte Maintenance Kit
STEMdiff™ Cardiomyocyte maintenance kit, 1 KIT
CAS Number:  
Price
2 249,00 kr
Log in for availability
1 * 1 KIT
Product Details & Documents
Medium for long-term maintenance of human PSC-derived cardiomyocytes.
STEMdiff™ Cardiomyocyte Maintenance Kit can be used for long-term maintenance of human pluripotent stem cell (PSC)-derived cardiomyocytes for one month or longer. The functional capacity of hPSC-derived cardiomyocytes is retained for downstream applications and analysis. This kit is compatible with cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Cardiomyocyte maintenance kit
Specifications