Order Entry
Sweden
Orders LinkContactUsLinkComponent
ITS Supplement-B
ITS Supplement-B
ITS supplement-B, 5 mL
Catalog # STMC07155
Marketsource
ITS Supplement-B
Catalog # STMC07155
Marketsource
CAS Number:  
ITS Supplement-B
ITS Supplement-B
ITS supplement-B, 5 mL
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 5 mL
Product Details & Documents
For neural differentiation of mouse ES and iPS cells.
ITS Supplement-B is a medium supplement containing insulin, apo-transferrin (iron-poor), and selenium in phosphate-buffered saline. It is recommended for use in the in vitro differentiation of mouse embryonic stem (ES) cells and mouse induced pluripotent stem (iPS) cells to neural cell types.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    5 mL
  • Description:
    ITS supplement-B