Order Entry
Sweden
ContactUsLinkComponent
 
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95% (dye content)
  252883-1G
 :  
undefined
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 95% (dye content)
  252883-1G
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance (Colour):
    Purple
  • Appearance (Form), Powder, Crystals, Crystalline Powder and/or Chunks:
    Conforms to Requirements
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength In Toluene:
    421 - 427 nm
  • Extinction Coefficient:
    ≥350000
  • Carbon:
    73.7 - 83.2%
  • Nitrogen:
    7.2 - 8.2%
  • Purity (HPLC):
    ≥94.0%

 

 

 :
 :734.86 g/mol
 :M-Clarity™ quality level = MQ100
 :Room Temperature
 :22112-78-3
 :C₄₈H₃₈N₄O₄
 :MFCD00012072