Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 700)
Supplier: AAT BIOQUEST
Certificates
About this item
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB101910J0
- AATB101910J1
- Antigen symbol:
- CD19
- CD19
- Antigen name:
- CD19 antigen
- CD19 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 700
- iFluor® 700
- Clone:
- SJ25C1
- SJ25C1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20