Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (APC (Allophycocyanin)/iFluor® 700)
Supplier: AAT Bioquest
Certificates
About this item
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: APC (Allophycocyanin)/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB100331F2
- AATB100331F0
- AATB100331F1
- Antigen symbol:
- CD3e
- CD3e
- CD3e
- Antigen name:
- CD3e antigen
- CD3e antigen
- CD3e antigen
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/iFluor® 700
- APC (Allophycocyanin)/iFluor® 700
- APC (Allophycocyanin)/iFluor® 700
- Clone:
- SK7
- SK7
- SK7
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
- 20
Specifications
Frequently Bought Together