Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ Violet 540)
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- AATB10032120
- AATB10032121
- Antigen symbol:
- CD3e
- CD3e
- Antigen name:
- CD3e antigen
- CD3e antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet 540
- mFluor™ Violet 540
- Clone:
- UCHT1
- UCHT1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:
- 20
- 20
Frequently Bought Together