Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (mFluor™ Violet 540)
Supplier: AAT Bioquest
Certificates
About this item
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB10021120
- AATB10021121
- Antigen symbol:
- CD2
- CD2
- Antigen name:
- CD2 antigen
- CD2 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ Violet 540
- mFluor™ Violet 540
- Clone:
- RPA-2.10
- RPA-2.10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together