Anti-CD38 Mouse Monoclonal Antibody [clone: HB7] (iFluor® 700)
Catalog # AATB103820J0
Supplier: AAT Bioquest
Specifications
- Antibody type:Primary
- Antigen name:CD38 antigen
- Antigen symbol:CD38
- Clonality:Monoclonal
- Clone:HB7
- Conjugation:iFluor® 700
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:lgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Storage temperature:2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 Tests
Specifications
About this item
HB7 is an anti-human monoclonal antibody that recognises the CD38 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human