Anti-SPDYE1 Rabbit Polyclonal Antibody
Catalog # PRSI55-768
Leverantör: ProSci Inc.
New Product
Specifikationer
- Antibody type:Primary
- Antigen name:Speedy protein E1
- Antigen symbol:SPDYE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino Acid Range = 242-271
- Form:Liquid
- Gene ID:Q8NFV5
- Antigen synonyms:SPDYE1
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:41 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SPDYE1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 242-271 amino acids from the C-terminal region of human SPDYE1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifikationer
Om denna produkt
This gene is located at chromosome 7p13 which is close to the Williams Beuren syndrome chromosome region 7q11.23. [provided by RefSeq].
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: SPDYE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human