- Antibody type:Primary
- Antigen name:Dictyoglomus thermophilum amylase gene
- Antigen symbol:Growth Hormone
- Clonality:Monoclonal
- Clone:GRHP1-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Format:Purified Antibody
- Form:Liquid
- Gene ID:X07896
- Antigen synonyms:Growth Hormone
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:A human partial recombinant protein was used as the immunogen for this Growth Hormone antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Pituitary growth hormone (GH) plays a crucial role in stimulating and controlling the growth, metabolism and differentiation of many mammalian cell types by modulating the synthesis of multiple mRNA species. These effects are mediated by the binding of GH to its membrane-bound receptor, GHR, and involve a phosphorylation cascade that results in the modulation of numerous signaling pathways. GH is synthesised by acidophilic or somatotropic cells of the anterior pituitary gland. Anti-GH is a useful marker in classification of pituitary tumors and the study of pituitary disease (acromegaly).
Flow Cytometry: 0,5 - 1 µg/10^6 cells IF: 1 - 2 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Growth Hormone antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate Buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone: GRHP1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human