Specifikationer
- Antibody type:Primary
- Antigen name:minichromosome maintenance complex component 7
- Antigen symbol:MCM7
- Clonality:Monoclonal
- Clone:MCM7/1467
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P33993
- Antigen synonyms:MCM7
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:A human partial recombinant protein corresponding to amino acids 195-319 was used as the immunogen for the MCM7 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifikationer
Om denna produkt
MCM7 is one of the highly conserved mini-chromosome maintenance proteins (MCM) that is essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumor suppressor protein RB1/RB.
Immunofluorescence: 0,5-1 µg/ml Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunohistochemistry (FFPE): 1 - 2 µg/ml for 30 min at RT Prediluted IHC only format: incubate for 30 min at RT (1) Titering of the MCM7 antibody may be required for optimal performance.
1. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MCM7
Clonality: Monoclonal
Clone: MCM7/1467
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human