- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU Antibody [BRD494]
- Clonality:Monoclonal
- Clone:BRD494
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Format:Purified Antibody
- Form:Liquid
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:BrdU / Bromodeoxyuridine conjugated to KLH was used as the immunogen for this antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to Bromodeoxyuridine/BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Bromodeoxyuridine/BrdU antibody to be titered up or down for optimal performance.
1. For staining of formalin-fixed tissues, incubate sections in 4N HCl for 30 minutes at RT followed by digestion with trypsin at 1mg/ml PBS, 10 min at 37oC.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: BrdU Antibody [BRD494]
Clonality: Monoclonal
Clone: BRD494
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species