Specifications
- Antibody type:Primary
- Antigen name:alpha-fetoprotein
- Antigen symbol:AFP
- Clonality:Monoclonal
- Clone:C3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Pig,Monkey,Human,Dog
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P02771
- Antigen synonyms:AFP
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Alpha fetoprotein purified from serum of a hepatoma patient was used as the immunogen for this AFP antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This antibody recognises an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as Alpha fetoprotein (AFP). The AFP antibody is highly specific and shows no cross-reaction with other oncofetal antigens or serum albumin. Alpha fetoprotein is normally synthesised in the liver, intestinal tract, and yolk sac of the fetus. AFP antibody has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml IHC(FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the AFP antibody to be titered up or down for optimal performance.
1. No special pretreatment is required for the immunohistochemical staining of formalin-fixed, paraffin-embedded tissues.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Monkey, Dog, Pig