Anti-DNASE1L1 Rabbit Polyclonal Antibody
PRSI14-819
- Antibody type:Primary
- Antigen name:Deoxyribonuclease-1-like 1
- Antigen symbol:DNASE1L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1774
- Antigen synonyms:DNASE1L1|XIB|DNase X|3121-|Deoxyribonuclease-1-like 1|DNAS1L1|Muscle-specific DNase I-like|DNase I-like 1|Deoxyribonuclease I-like 1|DNL1L
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-302 of human DNASE1L1 (NP_006721.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: DNASE1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat