Anti-UBA1 Rabbit Polyclonal Antibody
PRSI14-377
New Product
- Antibody type:Primary
- Antigen name:Ubiquitin-like modifier-activating enzyme 1
- Antigen symbol:UBA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:7317
- Antigen synonyms:A1S9T|UBE1|UBA1|Protein A1S9|Ubiquitin-like modifier-activating enzyme 1|Ubiquitin-activating enzyme E1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human UBA1 (NP_695012.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: UBA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat