Anti-BYSL Rabbit Polyclonal Antibody
Catalog # PRSI14-219
Leverantör: ProSci Inc.
New Product
Specifikationer
- Antibody type:Primary
- Antigen name:bystin like
- Antigen symbol:BYSL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:705
- Antigen synonyms:drosophila|bystin|homolog-like|Enp1|BYSTIN|by the ribosomal protein s6 gene
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 120-280 of human BYSL (NP_004044.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: BYSL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat