Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 860)
 
undefined
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 860)

 

  • Catalog No:
  • AATB101610R1
  • AATB101610R0
  • Antigensymbol:
  • CD16a
  • CD16a
  • Antikroppsnamn:
  • CD16a antigen
  • CD16a antigen
  • Antikroppstyp:
  • Primär
  • Primär
  • Klonalitet:
  • Monoclonal
  • Monoclonal
  • Konjugation:
  • iFluor® 860
  • iFluor® 860
  • Klon:
  • 3G8
  • 3G8
  • Reaktivitet:
  • Human
  • Human
  • Värd:
  • Mouse
  • Mouse
  • ImmunoFluorescens:
  • Yes
  • Yes
  • Flödescytometri:
  • Yes
  • Yes
  • Rening:
  • Affinity chromatography
  • Affinity chromatography
  • Korsad adsorption:
  • No
  • No
  • Förvaringstemperatur:
  • 2...8 °C with minimised light exposure. Do not freeze.
  • 2...8 °C with minimised light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20