Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (iFluor® 450)
Catalog # AATB10454040
Supplier: AAT Bioquest
Specifications
- Antibody type:Primary
- Antigen name:CD45 antigen
- Antigen symbol:CD45
- Clonality:Monoclonal
- Clone:2D1
- Conjugation:iFluor® 450
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Cross adsorption:No
- Storage temperature:2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:20
- Purification:Affinity chromatography
- Pack type:Vial
- Pk:100 Tests
Specifications
About this item
2D1 is an anti-human monoclonal antibody that recognises the CD45 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2D1
Conjugation: iFluor® 450
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human