Anti-G3BP2 Rabbit Polyclonal Antibody
Catalog # PRSI19-981
Supplier: ProSci Inc.
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:G3BP stress granule assembly factor 2
- Antigensymbol:G3BP2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:9908
- Antigen synonymer:G3BP-2|GTPase activating protein (SH3 domain) binding protein 2|GAP SH3 domain-binding protein 2|Ras-GTPase activating protein SH3 domain-binding protein 2|ras GTPase-activating protein-binding protein 2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Förvaringstemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Koncentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 140-280 of human G3BP2 (NP_987100.1).
- Rening:Affinity purification
- Förpackningstyp:Vial
- Förp.:50 µl
Specifications
About this item
Type: Primary
Antigen: G3BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat