Anti-GOLGA2 Rabbit Polyclonal Antibody
PRSI19-604
- Antibody type:Primary
- Antigen name:Golgin A2
- Antigen symbol:GOLGA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2801
- Antigen synonyms:GM13golgin subfamily A member 2|GM130 autoantigen|golgin subfamily a|Golgi matrix protein GM13SY11 protein|130 kDa cis-Golgi matrix protein|golgi autoantigen|golgin-95|2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human GM130 (NP_004477.3).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: GOLGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat