Anti-GOLT1B Rabbit Polyclonal Antibody
PRSI56-850
New Product
- Antibody type:Primary
- Antigen name:Vesicle transport protein GOT1B
- Antigen symbol:GOLT1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Epitope:Amino acid range = 108-136
- Form:Liquid
- Gene ID:Q9Y3E0
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:15 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GOT1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 108-136 amino acids from the C-terminal region of human GOT1B
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
GOT1B may be involved in fusion of ER-derived transport vesicles with the Golgi complex.
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: GOLT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse