Anti-SMARCE1 Rabbit Polyclonal Antibody
PRSI57-670
New Product
- Antibody type:Primary
- Antigen name:SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1
- Antigen symbol:SMARCE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:307 - 335 aa
- Form:Liquid
- Gene ID:6605
- Modification:None
- Molecular weight:47 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SMARCE1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 307-335 amino acids from the C-terminal region of human SMARCE1
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: SMARCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 307-335
Host: Rabbit
Isotype: Ig
Reactivity: Human