Anti-GATA1 Rabbit Polyclonal Antibody
Catalog # PRSI13-181
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:GATA binding protein 1 (globin transcription factor 1)
- Antigen symbol:GATA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:2623
- Antigen synonyms:ERYF1|NFE1|GF-1|GF1|GATA1|XLANP|XLTT|NF-E1|XLTDA|GATA-1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human GATA1
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GATA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat