Anti-DI-17E6 Recombinant Antibody
PRSI10-653
New Product
- Antibody type:Primary
- Clonality:Recombinant
- Conjugation:Unconjugated
- Antigen synonyms:TRU-016
- Modification:None
- Storage buffer:PBS buffer pH 7,5
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Humanised/CD37/TSPAN26 [Homo sapiens]
- Purification:>95%
- Pack type:Vial
- Pk:0,1 mg
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: VH-V-kappa-CH2-CH3
Reactivity: