Anti-PATE3 Rabbit Polyclonal Antibody
PRSI18-571
New Product
- Antibody type:Primary
- Antigen name:Prostate and testis expressed 3
- Antigen symbol:PATE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:100169851
- Antigen synonyms:HEL-127|prostate and testis expressed protein 3|PATE-like protein DJ|acrosomal vesicle protein HEL-127|secreted TFP/Ly-6/uPAR protein PATE-DJ|PATE-DJ
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human PATE3 (NP_001123355.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PATE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat