Anti-CENPJ Rabbit Polyclonal Antibody
PRSI23-708
- Antibody type:Primary
- Antigen name:centromere protein J
- Antigen symbol:CENPJ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:55835
- Antigen synonyms:LYST-interacting protein LIP7|LAG-3-associated protein|centromere protein J|Sas-4|CPAP|SASS4|LYST-interacting protein 1|LYST-interacting protein LIP1|LAP|BM032|MCPH6|SCKL4|Spindle assembly abnormal 4|centrosomal P4.1-associated protein|CENP-J|LIP1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1069-1338 of human CENPJ (NP_060921.3)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CENPJ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat