Anti-Cmklr1 Recombinant Antibody [Clone: BZ194]
Catalog # PRSI24-606
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:chemokine-like receptor 1
- Antigen symbol:Cmklr1
- Clonality:Recombinant
- Clone:BZ194
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:14747
- Antigen synonyms:DEZ|Dez|Gpcr27|Chemerin receptor|G-protein coupled receptor DEZ|ChemR23|Resolvin E1 receptor|MGC126105|RVER1|CHEMERINR|orphan G-protein coupled receptor|chemokine receptor-like 1|G-protein coupled receptor ChemR23|MGC126106|Chemokine-like receptor 1|CMKLR1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Wistar Furth rats with the mCMKLR1 peptide/KLH conjugate. The immunizing amino-terminal mCMKLR1 peptide with the sequence NH2-DSGIYDDEYSDGFGYFVDLEEASPWC-COOH, corresponding to residues residues 8–32 of mCMKLR1, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH)
- Tested applications:Functional assays
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: chemokine-like receptor 1
Clonality: Recombinant
Clone: BZ194
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse