Anti-MHC Class 1 Recombinant Antibody [Clone: R1-21.2]
Catalog # PRSI24-544
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:R1-21.2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:H-2|MHC-I|R1.21.2|MHC Class I
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Lewis 1F rats with mouse CH-1 lymphoma cells. Spleen cells from immunised rats were fused with X-63Ag8-653 myeloma cells to generate hybridomas
- Tested applications:Radioimmunoassay
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: MHC Class 1
Clonality: Recombinant
Clone: R1-21.2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Mouse