Anti-VEGFA Recombinant Antibody [Clone: Bevacizumab]
PRSI24-482
New Product
- Antibody type:Primary
- Antigen name:HGNC:HGNC:12680
- Antigen symbol:VEGFA
- Clonality:Recombinant
- Clone:Bevacizumab
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:7422
- Antigen synonyms:VEGFA|VEGF-A|Vascular Permeability Factor|VPF|ascular Endothelial Growth Factor
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human VEGF
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: HGNC:HGNC:12680
Clonality: Recombinant
Clone: Bevacizumab
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human