Anti-CD19 Recombinant Antibody [Clone: FMC63]
Catalog # PRSI24-434
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:HGNC:HGNC:1633
- Antigen symbol:CD19
- Clonality:Recombinant
- Clone:FMC63
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:930
- Antigen synonyms:Leu12|B-lymphocyte antigen CD19|Differentiation antigen CD19 T-cell surface antigen Leu-12|CD19|B-lymphocyte surface antigen B4
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human prolymphocytic leukaemia cell line JVM3
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: HGNC:HGNC:1633
Clonality: Recombinant
Clone: FMC63
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human