Order Entry
Sweden
ContactUsLinkComponent
Phenolphthalein 0.5% in aqueous solution
Phenolphthalein 0.5% in aqueous solution
Catalog # HACH16253
Supplier:  Hach
CAS Number:  
undefined
Phenolphthalein 0.5% in aqueous solution
Catalog # HACH16253
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :318.33 g/mol
Storage Temperature:Ambient
EINECS:201-004-7
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913