Order Entry
Sweden
ContactUsLinkComponent
Colophony (Rosin gum)
Irritant
Colophony (Rosin gum)
Certificates
undefined
Irritant
Colophony (Rosin gum)
Certificates

Specification Test Results

  • Appearance:
    Almost white to yellow to orange Powder or flakes or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 83 °C
  • Acid value:
    156 to 182 mg KOH/g
  • Uniformity coefficient:
    ≤1.8
  • Moisture holding capacity:
    53 to 58 % (H+)
  • Total volume capacity:
    ≥1.8 eq/l (H+)

Specification Test Results

Related Information