Order Entry
Sweden
ContactUsLinkComponent
Hydrogen peroxide 10% stabilised
Hydrogen peroxide 10% stabilised
Catalog # LBSE790841
Supplier:  LABSERVICE
CAS Number:  
Hydrogen peroxide 10% stabilised
Catalog # LBSE790841
Supplier:  LABSERVICE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Boiling Pt:150.2 °C (1013 hPa)
Melting Pt:–33 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:5.1,III
 :34.01 g/mol
EINECS:231-765-0
UN:2984
 :GHS05
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333