Order Entry
Sweden
ContactUsLinkComponent
Rhodamine B
Rhodamine B
Catalog # EHERC16813550
Supplier:  EHRENSTORFER
CAS Number:  
Rhodamine B
Catalog # EHERC16813550
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :479.02 g/mol
Merck Index:13,08266
Melting Pt:199…201 °C
Density:1.31 g/cm³ (20 °C)
EINECS:201-383-9
 :GHS05
Storage Temperature:Ambient
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931