Order Entry
Sweden
ContactUsLinkComponent
Eriochrome Black T
Eriochrome Black T
Catalog # MOLE51499212-25G
Supplier:  Molekula
CAS Number:  
Eriochrome Black T
Catalog # MOLE51499212-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:13,03702
ADR:9,III
 :461.39 g/mol
EINECS:217-250-3
UN:3077
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:1787-61-7
Formula:C₂₀H₁₂N₃NaO₇S
MDL Number:MFCD00003935