Eriochrome Black T
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1787-61-7
:
Formula:C₂₀H₁₂N₃NaO₇S
MDL Number:MFCD00003935
Merck Index:14,3667
ADR:9,III
:461.39 g/mol
EINECS:217-250-3
UN:3077
:GHS07
:GHS09
Storage Temperature:Ambient