Order Entry
Sweden
ContactUsLinkComponent
Bromophenol blue sodium salt
Bromophenol blue sodium salt
Catalog # SHETIB74040
Supplier:  IBI SCIENTIFIC
CAS Number:  
Bromophenol blue sodium salt
Catalog # SHETIB74040
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :691.95 g/mol
Storage Temperature:Ambient
EINECS:252-170-2
CAS Number:34725-61-6
 :62625-28-9
Formula:C₁₉H₉Br₄NaO₅S
MDL Number:MFCD00013793