


Isradipine ≥98%
Catalog # ENZOBMLCA2130010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
Specification Test Results
About this item
:
:371.39 g/mol
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:75695-93-1
Formula:C₁₉H₂₁N₃O₅
MDL Number:MFCD00153820