Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Isoquinolinediol
1,5-Isoquinolinediol ≥98%
1,5-Isoquinolinediol ≥98%
1,5-Isoquinolinediol ≥98%
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
1,5-Isoquinolinediol ≥98%
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
5154-02-9
:
Formula:
C₉H₇NO₂
MDL Number:
MFCD00006900
:
161.16 g/mol