Vitamin K1 (phytomenadione, phylloquinone)
Catalog # ENZOALX460027G005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:450.6958 g/mol
EINECS:201-564-2
CAS Number:84-80-0
:81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063