(-)-Arctiin (from Arctium lappa) ≥95% (by HPLC)
Catalog # ENZOALX350318M025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:534.6 g/mol
Storage Temperature:Freezer
CAS Number:20362-31-6
Formula:C₂₇H₃₄O₁₁
MDL Number:MFCD00870630