Order Entry
Sweden
ContactUsLinkComponent
Panepoxydone (from Lentinus crinitus) ≥95% (by HPLC, MS)
Panepoxydone (from Lentinus crinitus) ≥95% (by HPLC, MS)
Catalog # ENZOALX350109MC05
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Panepoxydone (from Lentinus crinitus) ≥95% (by HPLC, MS)
Catalog # ENZOALX350109MC05
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Seq:
    2035

Specifications

About this item

 :
 :210.2 g/mol
Storage Temperature:Freezer
CAS Number:31298-54-1
Formula:C₁₁H₁₄O₄