Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98%
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98%
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98%
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98%
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
119139-23-0
:
Formula:
C20H13N3O2
MDL Number:
MFCD00236432
:
327.3361 g/mol