Order Entry
Sweden
ContactUsLinkComponent
Di-2-ANEPEQ (JPW 1114) ≥90% (by HPLC), CELLESTIAL® membrane stain, for histology
Di-2-ANEPEQ (JPW 1114) ≥90% (by HPLC), CELLESTIAL® membrane stain, for histology
Catalog # ENZOENZ52201
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Di-2-ANEPEQ (JPW 1114) ≥90% (by HPLC), CELLESTIAL® membrane stain, for histology
Catalog # ENZOENZ52201
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :549.384 g/mol
Storage Temperature:Freezer
Formula:C₂₆H₃₅Br₂N₃