0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
JC-10
JC-10 ≥95% (by HPLC), Ultrapure
JC-10 ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52305
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
JC-10 ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52305
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
583.34 g/mol
Storage Temperature:
Freezer
Product Details