Order Entry
Sweden
ContactUsLinkComponent
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)
Catalog # ENZOBMLFR1130250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)
Catalog # ENZOBMLFR1130250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :424.9 g/mol
CAS Number:35369-53-0
Formula:C₂₀H₂₄N₂O₆·HCl·xH₂O
MDL Number:MFCD04113603