Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
GDC-0941
GDC-0941 ≥99% (by HPLC)
GDC-0941 ≥99% (by HPLC)
GDC-0941 ≥99% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
GDC-0941 ≥99% (by HPLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
957054-30-7
:
Formula:
C₂₃H₂₇N₇O₃S₂
MDL Number:
MFCD11616196
:
513.6 g/mol